3-(3-Fluorobenzyl)piperidine
Lieferant: Apollo Scientific
PC404526-1G
APOSPC404526-1GEA
467
EUR
APOSPC404526-1G
3-(3-Fluorobenzyl)piperidine
3-(3-Fluorobenzyl)piperidine
Formel:
C₁₂H₁₆FN Molecular Weight: 193,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863602 CAS-Nummer: 795261-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...