3-(3-Trifluoromethyl-benzyl)-piperidine
Lieferant: Apollo Scientific
PC404527-1G
APOSPC404527-1GEA
467
EUR
APOSPC404527-1G
3-(3-Trifluoromethyl-benzyl)-piperidine
3-(3-Trifluoromethyl-benzyl)-piperidine
Formel:
C₁₃H₁₆F₃N Molecular Weight: 243,268 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863603 CAS-Nummer: 625454-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...