3-(5-Fluoro-2-methoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC404528-1G
APOSPC404528-1GEA 219 EUR
APOSPC404528-1G
3-(5-Fluoro-2-methoxy-benzyl)-piperidine hydrochloride
3-(5-Fluoro-2-methoxy-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₈FNO·ClH
Molecular Weight: 259,747 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669682
CAS-Nummer: 1171433-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR