3-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC404529-1G
APOSPC404529-1GEA 185 EUR
APOSPC404529-1G
3-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
3-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₈FNO·ClH
Molecular Weight: 259,747 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08751317
CAS-Nummer: 1172742-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR