3-(3-Fluoro-4-methyl-benzyl)-piperidine

Lieferant: Apollo Scientific

PC404530-1G
APOSPC404530-1GEA 499 EUR
APOSPC404530-1G
3-(3-Fluoro-4-methyl-benzyl)-piperidine
3-(3-Fluoro-4-methyl-benzyl)-piperidine
Formel: C₁₃H₁₈FN
Molecular Weight: 207,287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863617
CAS-Nummer: 955288-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR