3-(4-Fluoro-2-methyl-benzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
PC404531-1G
APOSPC404531-1GEA
185
EUR
APOSPC404531-1G
3-(4-Fluoro-2-methyl-benzyl)-piperidine hydrochloride
3-(4-Fluoro-2-methyl-benzyl)-piperidine hydrochloride
Formel:
C₁₃H₁₈FN·ClH Molecular Weight: 243,748 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669684 CAS-Nummer: 1172955-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...