2-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC404532-1G
APOSPC404532-1GEA 433 EUR
APOSPC404532-1G
2-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
2-(3-Fluoro-4-methoxy-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₈FNO·ClH
Molecular Weight: 259,747 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669729
CAS-Nummer: 1170364-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR