2-(3-Fluoro-4-methyl-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC404533-1G
APOSPC404533-1GEA 161 EUR
APOSPC404533-1G
2-(3-Fluoro-4-methyl-benzyl)-piperidine hydrochloride
2-(3-Fluoro-4-methyl-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₈FN·ClH
Molecular Weight: 243,748 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669730
CAS-Nummer: 1172829-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR