2-(3-Fluoro-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC404534-1G
APOSPC404534-1GEA 433 EUR
APOSPC404534-1G
2-(3-Fluoro-benzyl)-piperidine hydrochloride
2-(3-Fluoro-benzyl)-piperidine hydrochloride
Formel: C₁₂H₁₆FN·ClH
Molecular Weight: 229,721 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669731
CAS-Nummer: 1172356-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR