2-(3-Trifluoromethyl-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

PC404535-1G
APOSPC404535-1GEA 433 EUR
APOSPC404535-1G
2-(3-Trifluoromethyl-benzyl)-piperidine hydrochloride
2-(3-Trifluoromethyl-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₆F₃N·ClH
Molecular Weight: 279,729 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669732
CAS-Nummer: 782504-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR