2-(4-Fluoro-benzyl)-piperidine

Lieferant: Apollo Scientific

PC404536-1G
APOSPC404536-1GEA 433 EUR
APOSPC404536-1G
2-(4-Fluoro-benzyl)-piperidine
2-(4-Fluoro-benzyl)-piperidine
Formel: C₁₂H₁₆FN
Molecular Weight: 193,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663624
CAS-Nummer: 67157-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR