(4-Fluorophenyl)(piperidin-4-yl)methanol

Lieferant: Apollo Scientific

PC404537-1G
APOSPC404537-1GEA 441 EUR
APOSPC404537-1G
(4-Fluorophenyl)(piperidin-4-yl)methanol
(4-Fluorophenyl)(piperidin-4-yl)methanol
Formel: C₁₂H₁₆FNO
Molecular Weight: 209,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06809667
CAS-Nummer: 54924-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR