1-[4-(Trifluoromethoxy)phenyl]cyclopropanecarboxylic acid

Lieferant: Apollo Scientific
Achtung

PC404538-1G PC404538-250MG
APOSPC404538-1GEA 0 EUR
APOSPC404538-1G APOSPC404538-250MG
1-[4-(Trifluoromethoxy)phenyl]cyclopropanecarboxylic acid
1-[4-(Trifluoromethoxy)phenyl]cyclopropanecarboxylic acid
Formel: C₁₁H₉F₃O₃
Schmelzpunkt: 104…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11037007
CAS-Nummer: 936727-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR