(1-(4-(Trifluoromethoxy)phenyl)cyclopropyl)methanamine hydrochloride

Lieferant: Apollo Scientific

PC404541-1G
APOSPC404541-1GEA 401 EUR
APOSPC404541-1G
(1-(4-(Trifluoromethoxy)phenyl)cyclopropyl)methanamine hydrochloride
(1-(4-(Trifluoromethoxy)phenyl)cyclopropyl)methanamine hydrochloride
Formel: C₁₁H₁₂F₃NO·ClH
Molecular Weight: 267,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619673
CAS-Nummer: 1209685-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR