(1-(4-Fluorophenyl)cyclopropyl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC404542-1G
APOSPC404542-1GEA 401 EUR
APOSPC404542-1G
(1-(4-Fluorophenyl)cyclopropyl)methanamine hydrochloride
(1-(4-Fluorophenyl)cyclopropyl)methanamine hydrochloride
Formel: C₁₀H₁₂FN·ClH
Molecular Weight: 201,668 g/mol
Schmelzpunkt: 192…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09864782
CAS-Nummer: 1209726-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR