2-Bromo-3,3,3-trifluoro-1-(4-methylpiperazin-1-yl)propan-1-one

Lieferant: Apollo Scientific

PC404543-1G
APOSPC404543-1GEA 219 EUR
APOSPC404543-1G
2-Bromo-3,3,3-trifluoro-1-(4-methylpiperazin-1-yl)propan-1-one
2-Bromo-3,3,3-trifluoro-1-(4-methylpiperazin-1-yl)propan-1-one
Formel: C₈H₁₂BrF₃N₂O
Molecular Weight: 289,093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997950
CAS-Nummer: 1031927-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR