(R*,R*)-(2,8-Bis-trifluoromethyl-quinolin-4-yl)-piperidin-2-yl-methanol hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-[2,8-Bis(trifluoromethyl)-4-quinolinyl][(2R)-2-piperidinyl]methanol hydrochloride
PC404547-250MG
APOSPC404547-250MGEA
48
EUR
APOSPC404547-250MG
(R*,R*)-(2,8-Bis-trifluoromethyl-quinolin-4-yl)-piperidin-2-yl-methanol hydrochloride
(R*,R*)-(2,8-Bis-trifluoromethyl-quinolin-4-yl)-piperidin-2-yl-methanol hydrochloride
Formel:
C₁₇H₁₆F₆N₂O·ClH Molecular Weight: 414,773 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437514 CAS-Nummer: 58560-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...