(2,8-Bis-trifluoromethyl-quinolin-4-yl)-pyridin-2-yl-methanol

Lieferant: Apollo Scientific

PC404548-1G
APOSPC404548-1GEA 325 EUR
APOSPC404548-1G
(2,8-Bis-trifluoromethyl-quinolin-4-yl)-pyridin-2-yl-methanol
(2,8-Bis-trifluoromethyl-quinolin-4-yl)-pyridin-2-yl-methanol
Formel: C₁₇H₁₀F₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD05863552
CAS-Nummer: 68496-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR