6-(2,8-Bis-trifluoromethyl-quinolin-4-ylmethyl)-pyridin-3-ol

Lieferant: Apollo Scientific

PC404549-1G
APOSPC404549-1GEA 451 EUR
APOSPC404549-1G
6-(2,8-Bis-trifluoromethyl-quinolin-4-ylmethyl)-pyridin-3-ol
6-(2,8-Bis-trifluoromethyl-quinolin-4-ylmethyl)-pyridin-3-ol
Formel: C₁₇H₁₀F₆N₂O
Molecular Weight: 372,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863553
CAS-Nummer: 955315-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR