2,8-Bis(trifluoromethyl)-2'-(2-pyridyl)-4-quinolineacetonitrile

Lieferant: Apollo Scientific

PC404550-1G
APOSPC404550-1GEA 375 EUR
APOSPC404550-1G
2,8-Bis(trifluoromethyl)-2'-(2-pyridyl)-4-quinolineacetonitrile
2,8-Bis(trifluoromethyl)-2'-(2-pyridyl)-4-quinolineacetonitrile
Formel: C₁₈H₉F₆N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08437551
CAS-Nummer: 83012-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR