4-Hydrazino 7-trifluoromethyl-quinoline hydrochloride

Lieferant: Apollo Scientific

PC404551-1G
APOSPC404551-1GEA 335 EUR
APOSPC404551-1G
4-Hydrazino 7-trifluoromethyl-quinoline hydrochloride
4-Hydrazino 7-trifluoromethyl-quinoline hydrochloride
Formel: C₁₀H₈F₃N₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD02684138
CAS-Nummer: 88164-54-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR