4-Hydrazino 8-trifluoromethyl-quinoline hydrochloride

Lieferant: Apollo Scientific

PC404552-1G
APOSPC404552-1GEA 335 EUR
APOSPC404552-1G
4-Hydrazino 8-trifluoromethyl-quinoline hydrochloride
4-Hydrazino 8-trifluoromethyl-quinoline hydrochloride
Formel: C₁₀H₈F₃N₃·ClH
Molecular Weight: 263,647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703330
CAS-Nummer: 1171001-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR