2-[2,8-Bis(trifluoromethyl)-4-quinolyl]oxirane
Lieferant: Apollo Scientific
PC404553-1G
APOSPC404553-1GEA
516
EUR
APOSPC404553-1G
2-[2,8-Bis(trifluoromethyl)-4-quinolyl]oxirane
2-[2,8-Bis(trifluoromethyl)-4-quinolyl]oxirane
Formel:
C₁₃H₇F₆NO Molecular Weight: 307,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260425 CAS-Nummer: 57120-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...