2-Chlor-3-(chlormethyl)-6-fluorchinolin

Lieferant: Apollo Scientific

PC404554-1G
APOSPC404554-1GEA 393 EUR
APOSPC404554-1G
2-Chlor-3-(chlormethyl)-6-fluorchinolin
2-Chloro-3-(chloromethyl)-6-fluoroquinoline
Formel: C₁₀H₆Cl₂FN
Lagertemperatur: Raumtemperatur
CAS-Nummer: 948291-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR