2-Chloro-6-fluoroquinoline-3-carbonitrile

Lieferant: Apollo Scientific

PC404557-1G
APOSPC404557-1GEA 383 EUR
APOSPC404557-1G
2-Chloro-6-fluoroquinoline-3-carbonitrile
2-Chloro-6-fluoroquinoline-3-carbonitrile
Formel: C₁₀H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787672
CAS-Nummer: 948291-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR