2-Chloro-6-fluoroquinoline-3-carboxaldehyde

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-6-fluoroquinoline-3-carbaldehyde

PC404558-1G
APOSPC404558-1GEA 430 EUR
APOSPC404558-1G
2-Chloro-6-fluoroquinoline-3-carboxaldehyde
2-Chloro-6-fluoroquinoline-3-carboxaldehyde
Formel: C₁₀H₅ClFNO
Molecular Weight: 209,604 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06337019
CAS-Nummer: 749920-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR