6-Fluoroquinolin-4-ylamine
Lieferant: Apollo Scientific
Synonyme:
4-Amino-6-fluoroquinoline
PC404560-1G
PC404560-5G
APOSPC404560-1GEA
343
EUR
APOSPC404560-1G
APOSPC404560-5G
6-Fluoroquinolin-4-ylamine
6-Fluoroquinolin-4-ylamine
Formel:
C₉H₇FN₂ Molecular Weight: 162.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664759 CAS-Nummer: 874800-60-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...