6-Fluoroquinolin-4-ylamine

Lieferant: Apollo Scientific

Synonyme: 4-Amino-6-fluoroquinoline

PC404560-1G PC404560-5G
APOSPC404560-1GEA 343 EUR
APOSPC404560-1G APOSPC404560-5G
6-Fluoroquinolin-4-ylamine
6-Fluoroquinolin-4-ylamine
Formel: C₉H₇FN₂
Molecular Weight: 162.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664759
CAS-Nummer: 874800-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR