4-Amino-7-fluoro-2-methylquinoline
Lieferant: Apollo Scientific
PC404561-1G
APOSPC404561-1GEA
0
EUR
APOSPC404561-1G
4-Amino-7-fluoro-2-methylquinoline
4-Amino-7-fluoro-2-methylquinoline
Formel:
C₁₀H₉FN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787722 CAS-Nummer: 948293-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...