4-Chlor-7-fluorchinolin
Lieferant: Apollo Scientific
|
Achtung
|
PC404563-1G
APOSPC404563-1GEA
62.5
EUR
APOSPC404563-1G
4-Chlor-7-fluorchinolin
4-Chloro-7-fluoroquinoline
Formel:
C₉H₅ClFN Molecular Weight: 181.5941 g/mol Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153094 CAS-Nummer: 391-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...