7-Fluor-2-methylchinolin-3-carbonsäure

Lieferant: Apollo Scientific

PC404564-1G
APOSPC404564-1GEA 383 EUR
APOSPC404564-1G
7-Fluor-2-methylchinolin-3-carbonsäure
7-Fluor-2-methylchinolin-3-carbonsäure
Formel: C₁₁H₈FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06254735
CAS-Nummer: 879361-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR