6-Fluoro-2-hydroxy-3-phenylquinoline

Lieferant: Apollo Scientific

PC404565-1G
APOSPC404565-1GEA 368 EUR
APOSPC404565-1G
6-Fluoro-2-hydroxy-3-phenylquinoline
6-Fluoro-2-hydroxy-3-phenylquinoline
Formel: C₁₅H₁₀FNO
Molecular Weight: 239,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997959
CAS-Nummer: 1031928-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR