2,8-Bis(trifluoromethyl)-4-vinylquinoline

Lieferant: Apollo Scientific
Achtung

PC404566-1G
APOSPC404566-1GEA 525 EUR
APOSPC404566-1G
2,8-Bis(trifluoromethyl)-4-vinylquinoline
2,8-Bis(trifluoromethyl)-4-vinylquinoline
Formel: C₁₃H₇F₆N
Molecular Weight: 291,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703428
CAS-Nummer: 1031928-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR