4-Brom-6-fluorchinolin
Lieferant: Apollo Scientific
PC404567-1G
APOSPC404567-1GEA
68.3
EUR
APOSPC404567-1G
4-Brom-6-fluorchinolin
4-Bromo-6-fluoroquinoline
Formel:
C₉H₅BrFN Molecular Weight: 226,0500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261094 CAS-Nummer: 661463-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...