4-Bromo-7-fluoroquinoline 95%

Lieferant: Apollo Scientific

PC404568-1G
APOSPC404568-1GEA 106 EUR
APOSPC404568-1G
4-Bromo-7-fluoroquinoline 95%
4-Bromo-7-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063190
CAS-Nummer: 1070879-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR