4-Bromo-6-trifluoromethylquinoline

Lieferant: Apollo Scientific

PC404569-1G
APOSPC404569-1GEA 69.6 EUR
APOSPC404569-1G
4-Bromo-6-trifluoromethylquinoline
4-Bromo-6-trifluoromethylquinoline
Formel: C₁₀H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD11505113
CAS-Nummer: 1070879-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR