4-Bromo-7-trifluoromethylquinoline
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-7-(trifluoromethyl)quinoline
PC404570-1G
APOSPC404570-1GEA
565
EUR
APOSPC404570-1G
4-Bromo-7-trifluoromethylquinoline
4-Bromo-7-trifluoromethylquinoline
Formel:
C₁₀H₅BrF₃N Molecular Weight: 276,053 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08063192 CAS-Nummer: 89446-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...