4-Bromo-8-trifluoromethylquinoline 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-8-(trifluoromethyl)quinoline

PC404571-1G
APOSPC404571-1GEA 139 EUR
APOSPC404571-1G
4-Bromo-8-trifluoromethylquinoline 95%
4-Bromo-8-trifluoromethylquinoline
Formel: C₁₀H₅BrF₃N
Molecular Weight: 276,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063211
CAS-Nummer: 260973-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR