4-Bromo-8-fluoro-2-methylquinoline

Lieferant: Apollo Scientific

PC404572-1G
APOSPC404572-1GEA 409 EUR
APOSPC404572-1G
4-Bromo-8-fluoro-2-methylquinoline
4-Bromo-8-fluoro-2-methylquinoline
Formel: C₁₀H₇BrFN
Molecular Weight: 240,072 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505134
CAS-Nummer: 1070879-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR