4-Bromo-2-methyl-8-trifluoromethylquinoline

Lieferant: Apollo Scientific

PC404573-1G
APOSPC404573-1GEA 375 EUR
APOSPC404573-1G
4-Bromo-2-methyl-8-trifluoromethylquinoline
4-Bromo-2-methyl-8-trifluoromethylquinoline
Formel: C₁₁H₇BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD11505142
CAS-Nummer: 1070879-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR