5,8-Difluor-4-chinolinol

Lieferant: Apollo Scientific

Synonyme: 5,8-Difluoro-4-hydroxyquinoline

PC404576-1G
APOSPC404576-1GEA 139 EUR
APOSPC404576-1G
5,8-Difluor-4-chinolinol
5,8-Difluor-4-chinolinol
Formel: C₉H₅F₂NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 874781-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR