7,8-Difluoro-4-hydroxyquinoline

Lieferant: Apollo Scientific

PC404577-1G
APOSPC404577-1GEA 351 EUR
APOSPC404577-1G
7,8-Difluoro-4-hydroxyquinoline
7,8-Difluoro-4-hydroxyquinoline
Formel: C₉H₅F₂NO
Molecular Weight: 181,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870786
CAS-Nummer: 1142193-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR