4-Amino-8-trifluoromethoxyquinoline

Lieferant: Apollo Scientific

PC404579-1G
APOSPC404579-1GEA 491 EUR
APOSPC404579-1G
4-Amino-8-trifluoromethoxyquinoline
4-Amino-8-trifluoromethoxyquinoline
Formel: C₁₀H₇F₃N₂O
Molecular Weight: 228,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059368
CAS-Nummer: 874880-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR