5,7-Difluoro-4-hydroxyquinoline
Lieferant: Apollo Scientific
PC404580-1G
APOSPC404580-1GEA
174
EUR
APOSPC404580-1G
5,7-Difluoro-4-hydroxyquinoline
5,7-Difluoro-4-hydroxyquinoline
Formel:
C₉H₅F₂NO Molecular Weight: 181,139 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06409136 CAS-Nummer: 874804-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...