4-Bromo-5,7-difluoroquinoline

Lieferant: Apollo Scientific

PC404581-1G
APOSPC404581-1GEA 517 EUR
APOSPC404581-1G
4-Bromo-5,7-difluoroquinoline
4-Bromo-5,7-difluoroquinoline
Formel: C₉H₄BrF₂N
Molecular Weight: 244,036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674965
CAS-Nummer: 1189107-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR