4-Chloro-7,8-difluoroquinoline

Lieferant: Apollo Scientific

PC404583-1G
APOSPC404583-1GEA 475 EUR
APOSPC404583-1G
4-Chloro-7,8-difluoroquinoline
4-Chloro-7,8-difluoroquinoline
Formel: C₉H₄ClF₂N
Molecular Weight: 199,585 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674973
CAS-Nummer: 1189105-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR