4-Amino-7,8-difluoroquinoline

Lieferant: Apollo Scientific

PC404585-1G
APOSPC404585-1GEA 540 EUR
APOSPC404585-1G
4-Amino-7,8-difluoroquinoline
4-Amino-7,8-difluoroquinoline
Formel: C₉H₆F₂N₂
Molecular Weight: 180.154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674976
CAS-Nummer: 1189107-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR