4-Bromo-2-methyl-6-trifluoromethoxyquinoline
Lieferant: Apollo Scientific
PC404586-1G
APOSPC404586-1GEA
607
EUR
APOSPC404586-1G
4-Bromo-2-methyl-6-trifluoromethoxyquinoline
4-Bromo-2-methyl-6-trifluoromethoxyquinoline
Formel:
C₁₁H₇BrF₃NO Molecular Weight: 306,079 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12675022 CAS-Nummer: 1189107-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...