5,7-Difluoro-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 5,7-Difluoro-4-hydroxy-2-methyl-quinoline

PC404587-1G
APOSPC404587-1GEA 223 EUR
APOSPC404587-1G
5,7-Difluoro-4-hydroxy-2-methylquinoline
5,7-Difluoro-4-hydroxy-2-methylquinoline
Formel: C₁₀H₇F₂NO
Molecular Weight: 195,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501289
CAS-Nummer: 288151-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR