4-Chloro-5,7-difluoro-2-methylquinoline

Lieferant: Apollo Scientific

PC404588-1G
APOSPC404588-1GEA 383 EUR
APOSPC404588-1G
4-Chloro-5,7-difluoro-2-methylquinoline
4-Chloro-5,7-difluoro-2-methylquinoline
Formel: C₁₀H₆ClF₂N
Molecular Weight: 213,611 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501279
CAS-Nummer: 288151-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR